Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAPK2
Entrez Gene ID
102169967
Gene Name
death associated protein kinase 2
Gene Aliases
-
Chromosome Location
Chr.10: 57001419 - 57133564 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054331.1 XP_017909820.1 3 - 4 440 61
GenBank mRNA JO584087.1 - 9 - 10 1016 61

View Full Product Details