Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC3A2
Entrez Gene ID
102187568
Gene Name
solute carrier family 3 member 2
Gene Aliases
-
Chromosome Location
Chr.29: 41488421 - 41519492 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699799.3 XP_005699856.1 5 - 6 1089 62
GenBank mRNA JO611079.2 - 5 - 6 1128 62
JO643663.2 - 5 - 6 952 62

View Full Product Details