Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TULP3
Entrez Gene ID
102170303
Gene Name
tubby like protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 105767749 - 105798009 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048629.1 XP_017904118.1 6 - 7 899 67
XM_018048630.1 XP_017904119.1 7 - 8 2257 67
XM_018048631.1 XP_017904120.1 6 - 7 2230 67
XM_018048632.1 XP_017904121.1 6 - 7 824 67
XM_018048633.1 XP_017904122.1 5 - 6 754 67
GenBank mRNA JO426864.1 - 6 - 7 940 67
KA339513.1 - 6 - 7 824 67

View Full Product Details