Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PANX3
Entrez Gene ID
102180016
Gene Name
pannexin 3
Gene Aliases
-
Chromosome Location
Chr.29: 27422712 - 27431582 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699573.3 XP_005699630.1 3 - 4 545 79

View Full Product Details