Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL5
Entrez Gene ID
102175198
Gene Name
synaptotagmin like 5
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 31768015 - 31883530 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700922.3 XP_005700979.1 4 - 5 906 61

View Full Product Details