Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRMPD3
Entrez Gene ID
102178802
Gene Name
FERM and PDZ domain containing 3
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189517: 35739953 - 35808095 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044100.1 XP_017899589.1 13 - 14 1480 58

Gene Symbol
LOC108634520
Entrez Gene ID
108634520
Gene Name
FERM and PDZ domain-containing protein 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189547: 66120 - 72337 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044449.1 XP_017899938.1 - 820 58

View Full Product Details