Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARGFX
Entrez Gene ID
102172199
Gene Name
arginine-fifty homeobox
Gene Aliases
-
Chromosome Location
Chr.1: 65757488 - 65768492 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005709423.2 XP_005709480.2 2 - 3 312 78

View Full Product Details