Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAWR
Entrez Gene ID
102186745
Gene Name
pro-apoptotic WT1 regulator
Gene Aliases
-
Chromosome Location
Chr.5: 8939100 - 9057056 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047593.1 XP_017903082.1 2 - 3 874 64
XM_018047594.1 XP_017903083.1 2 - 3 877 64

View Full Product Details