Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP298
Entrez Gene ID
102177277
Gene Name
cilia and flagella associated protein 298
Gene Aliases
C1H21orf59
Chromosome Location
Chr.1: 1384165 - 1397044 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674692.3 XP_005674749.2 5 - 6 1416 121
GenBank mRNA JO353076.2 - 2 - 3 143 121
JO415835.2 - 2 - 3 146 121
JO437008.2 - 5 - 6 912 121
JO504230.2 - 5 - 6 824 121
JO551203.2 - 2 - 3 121
JO594966.2 - 5 - 6 904 121
JO631207.2 - 5 - 6 866 121
KA321950.1 - 5 - 6 999 121

View Full Product Details