Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPOCD1
Entrez Gene ID
102172308
Gene Name
SPOC domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 14033186 - 14064006 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066346.1 XP_017921835.1 9 - 10 2050 71

View Full Product Details