Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC85A
Entrez Gene ID
102176429
Gene Name
coiled-coil domain containing 85A
Gene Aliases
-
Chromosome Location
Chr.11: 38443505 - 38665181 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055267.1 XP_017910756.1 1 - 2 1103 58
XM_018055269.1 XP_017910758.1 1 - 2 1103 58
XM_018055270.1 XP_017910759.1 1 - 2 1103 58

View Full Product Details