Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXN2
Entrez Gene ID
102174035
Gene Name
forkhead box N2
Gene Aliases
-
Chromosome Location
Chr.11: 30234553 - 30290610 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055206.1 XP_017910695.1 5 - 6 1114 81

View Full Product Details