Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFS1
Entrez Gene ID
102188574
Gene Name
wolframin ER transmembrane glycoprotein
Gene Aliases
-
Chromosome Location
Chr.6: 103268288 - 103299904 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049710.1 XP_017905199.1 6 - 7 850 54

View Full Product Details