Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUS3
Entrez Gene ID
102186838
Gene Name
pseudouridine synthase 3
Gene Aliases
-
Chromosome Location
Chr.29: 28591022 - 28601274 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699602.3 XP_005699659.2 4 - 5 1227 85
XM_005699604.3 XP_005699661.2 5 - 6 1342 85
XM_005699606.3 XP_005699663.2 4 - 5 1259 85
GenBank mRNA JO617380.2 - 1 - 2 193 85
KA346503.1 - 1 - 2 85

View Full Product Details