Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
IL4I1
Entrez Gene ID
102186899
Gene Name
interleukin 4 induced 1
Gene Aliases
-
Chromosome Location
Chr.18: 57244341 - 57249585 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062816.1 XP_017918305.1 4 - 5 561 89
XM_018062817.1 XP_017918306.1 4 - 5 422 89

View Full Product Details