Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICMT
Entrez Gene ID
102181140
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
-
Chromosome Location
Chr.16: 45640500 - 45650029 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060445.1 XP_017915934.1 4 - 5 745 68

View Full Product Details