Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARM1
Entrez Gene ID
102181283
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
-
Chromosome Location
Chr.6: 90373307 - 90505141 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049587.1 XP_017905076.1 3 - 4 1077 68

View Full Product Details