Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCH7
Entrez Gene ID
102188981
Gene Name
membrane associated ring-CH-type finger 7
Gene Aliases
-
Chromosome Location
Chr.2: 99549257 - 99596849 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676076.3 XP_005676133.1 6 - 7 1949 75
XM_013969446.2 XP_013824900.1 6 - 7 1950 75
XM_013969465.2 XP_013824919.1 6 - 7 1950 75
XM_013969466.2 XP_013824920.1 5 - 6 1797 75
XM_018063557.1 XP_017919046.1 6 - 7 1886 75
XM_018063558.1 XP_017919047.1 5 - 6 1797 75
XM_018063564.1 XP_017919053.1 5 - 6 1785 75
XM_018063568.1 XP_017919057.1 5 - 6 1706 75
GenBank mRNA KA319812.1 - 4 - 5 903 75

View Full Product Details