Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102175340
Entrez Gene ID
102175340
Gene Name
serine protease 42
Gene Aliases
PRSS42
Chromosome Location
Chr.22: 52700741 - 52705628 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038481.1 XP_017893970.1 2 - 3 592 59

View Full Product Details