Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA6
Entrez Gene ID
102188285
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Gene Aliases
-
Chromosome Location
Chr.27: 8098536 - 8103967 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041757.1 XP_017897246.1 3 - 4 242 61

View Full Product Details