Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPSM3
Entrez Gene ID
102191567
Gene Name
G protein signaling modulator 3
Gene Aliases
-
Chromosome Location
Chr.23: 22752455 - 22754579 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696532.3 XP_005696589.1 1 - 2 397 67
XM_005696533.3 XP_005696590.1 1 - 2 390 67
GenBank mRNA JO341606.2 - 3 - 4 1218 67
JO579847.1 - 1 - 2 436 67

View Full Product Details