Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARTN
Entrez Gene ID
106501979
Gene Name
artemin
Gene Aliases
-
Chromosome Location
Chr.3: 18642804 - 18646519 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043070.1 XP_017898559.1 1 - 2 2288 73

View Full Product Details