Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRSS57
Entrez Gene ID
102172436
Gene Name
serine protease 57
Gene Aliases
-
Chromosome Location
Chr.7: 67187979 - 67193164 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050506.1 XP_017905995.1 4 - 5 690 88

View Full Product Details