Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAKD
Entrez Gene ID
102187526
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
-
Chromosome Location
Chr.19: 44353495 - 44372968 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065258.1 XP_017920747.1 4 - 5 598 73
GenBank mRNA JO583351.1 - 4 - 5 633 73

View Full Product Details