Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFSF9
Entrez Gene ID
102188582
Gene Name
TNF superfamily member 9
Gene Aliases
-
Chromosome Location
Chr.7: 92527349 - 92531369 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050942.1 XP_017906431.1 1 - 2 413 72

View Full Product Details