Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSF5
Entrez Gene ID
102188079
Gene Name
heat shock transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.19: 9021773 - 9073129 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064237.1 XP_017919726.1 4 - 5 1621 99

View Full Product Details