Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSX1
Entrez Gene ID
102185375
Gene Name
visual system homeobox 1
Gene Aliases
-
Chromosome Location
Chr.13: 42035817 - 42044098 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056889.1 XP_017912378.1 3 - 4 697 59

View Full Product Details