Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM44
Entrez Gene ID
102182988
Gene Name
tripartite motif containing 44
Gene Aliases
-
Chromosome Location
Chr.15: 16635793 - 16748223 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690119.3 XP_005690176.3 2 - 3 1065 88
XM_018059237.1 XP_017914726.1 2 - 3 1065 88

View Full Product Details