Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPP4
Entrez Gene ID
102169156
Gene Name
phospholipid phosphatase 4
Gene Aliases
-
Chromosome Location
Chr.26: 11346974 - 11490959 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041752.1 XP_017897241.1 2 - 3 222 68

View Full Product Details