Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WAPL
Entrez Gene ID
100861086
Gene Name
WAPL cohesin release factor
Gene Aliases
WAPAL
Chromosome Location
Chr.28: 4487893 - 4565032 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699311.3 XP_005699368.1 15 - 16 3581 61
XM_018042447.1 XP_017897936.1 15 - 16 3568 61
GenBank mRNA JI861778.1 - 13 - 14 2965 61

View Full Product Details