Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK13
Entrez Gene ID
102180530
Gene Name
kallikrein related peptidase 13
Gene Aliases
-
Chromosome Location
Chr.18: 58112444 - 58122095 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692758.3 XP_005692815.1 5 - 6 799 74
GenBank mRNA KA352230.1 - 1 - 2 578 74

View Full Product Details