Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCBE1
Entrez Gene ID
102175176
Gene Name
collagen and calcium binding EGF domains 1
Gene Aliases
-
Chromosome Location
Chr.24: 58574649 - 58812785 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039646.1 XP_017895135.1 6 - 7 744 54
GenBank mRNA JO433355.2 - 6 - 7 674 54

View Full Product Details