Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD7
Entrez Gene ID
102187527
Gene Name
CD7 molecule
Gene Aliases
-
Chromosome Location
Chr.19: 49917649 - 49934714 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065405.1 XP_017920894.1 6 - 7 975 100

View Full Product Details