Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk7
Entrez Gene ID
100379283
Gene Name
potassium two pore domain channel subfamily K member 7
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176377: 6297683 - 6300234 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172762.1 NP_001166233.1 1 - 2 312 76
GenBank mRNA AY075099.1 - 1 - 2 312 76

View Full Product Details