The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_135992.4 | NP_609836.1 | 3 - 4 | 421 | 82 |
GenBank mRNA | BT128818.1 | - | 3 - 4 | 416 | 82 |