Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SREBP
Entrez Gene ID
40155
Gene Name
Sterol regulatory element binding protein
Gene Aliases
CG8522, Dmel\CG8522, Dmel_CG8522, HLH106, HlH106, SREBF1, Srebp, dSREBP, dSrebp, dsrebp, l(3)76BDw, srebp
Chromosome Location
Chr.3L: 19793935 - 19798979 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261283
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275135.1 NP_001262064.1 7 - 8 3317 69
NM_079442.3 NP_524166.2 6 - 7 3215 69
NM_168815.2 NP_730449.1 7 - 8 3238 69
NM_168816.2 NP_730450.1 7 - 8 3348 69
GenBank mRNA BT010285.1 - 7 - 8 3229 69

View Full Product Details