Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aldh-III
Entrez Gene ID
45398
Gene Name
Aldehyde dehydrogenase type III
Gene Aliases
ALDH, ALDH-III, AldhIII, CG11140, CT41571, Dhap, Dmel\CG11140, Dmel_CG11140, FALDH, l(2)03610
Chromosome Location
Chr.2R: 7466879 - 7477162 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010548
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273843.1 NP_001260772.1 7 - 8 1070 58
NM_001273844.1 NP_001260773.1 7 - 8 1197 58
NM_001273846.1 NP_001260775.1 7 - 8 1070 58
NM_136441.4 NP_610285.5 9 - 10 1536 58
NM_165526.3 NP_724560.1 6 - 7 1269 58
NM_165527.3 NP_724561.2 7 - 8 1108 58
NM_165528.3 NP_724562.2 9 - 10 2057 58
NM_165529.3 NP_724563.2 7 - 8 1016 58
NM_165530.3 NP_724564.2 7 - 8 1197 58
NM_165531.3 NP_724565.3 9 - 10 1412 58
NM_165532.3 NP_724566.3 9 - 10 1316 58
NM_206050.3 NP_995772.1 7 - 8 1070 58
GenBank mRNA BT003248.1 - 6 - 7 1278 58
BT003252.1 - 7 - 8 1165 58
BT044325.1 - 7 - 8 1069 58
BT126071.1 - 9 - 10 1563 58

View Full Product Details