Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mef2
Entrez Gene ID
36032
Gene Name
Myocyte enhancer factor 2
Gene Aliases
22.21, BEST:SD04091, C, CG1429, D-MEF2, D-Mef2, D-mef2, DMEF-2, DMEF2, DMedf2, DMef-2, DMef2, Dmef, Dmef-2, Dmef2, Dmel\CG1429, Dmel_CG1429, MEF-2, MEF2, Mef, Mef-2, SD04091, dMEF2, dMEf2, dMef2, dmef2, l(2)46CFr, mb247, mef, mef-2, mef2, mef2c
Chromosome Location
Chr.2R: 9913496 - 9958858 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011656
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273912.1 NP_001260841.1 5 - 6 2720 66
NM_001273913.2 NP_001260842.1 4 - 5 1394 66
NM_001382026.1 NP_001369068.1 3 - 4 1316 66
NM_001382027.1 NP_001369069.1 3 - 4 1316 66
NM_001382028.1 NP_001369070.1 3 - 4 1316 66
NM_001382029.1 NP_001369071.1 3 - 4 1316 66
NM_057670.5 NP_477018.1 3 - 4 1316 66
NM_057671.5 NP_477019.1 3 - 4 1316 66
NM_057672.5 NP_477020.1 4 - 5 1699 66
NM_057673.5 NP_477021.1 3 - 4 1316 66
NM_206067.4 NP_995789.1 4 - 5 1544 66
GenBank mRNA BT133393.1 - 3 - 4 1293 66
X83527.1 - - 1298 66

View Full Product Details