Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HBS1
Entrez Gene ID
117365
Gene Name
CG1898-PA
Gene Aliases
CG1898, Dmel\CG1898, Dmel_CG1898, Hbs1, hbs1
Chromosome Location
Chr.3L: 1862881 - 1865240 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0042712
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299977.1 NP_001286906.1 1 - 1 2313 127
NM_144472.4 NP_652729.2 1 - 1 2212 127
GenBank mRNA BT120040.1 - 1 - 1 2188 127

View Full Product Details