Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Obp50b
- Entrez Gene ID
-
246435
- Gene Name
- Odorant-binding protein 50b
- Gene Aliases
-
CG13940, CG30073, DmelObp50b, Dmel\CG30073, Dmel_CG30073
- Chromosome Location
-
Chr.2R: 14371186 - 14373006 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050073
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166051.3 |
NP_725386.1 |
1 - 2 |
267 |
62 |
GenBank mRNA |
BT088819.1 |
- |
1 - 2 |
267 |
62 |
- Gene Symbol
- Obp50c
- Entrez Gene ID
-
246434
- Gene Name
- Odorant-binding protein 50c
- Gene Aliases
-
CG30072, DmelObp50c, Dmel\CG30072, Dmel_CG30072
- Chromosome Location
-
Chr.2R: 14371186 - 14373006 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050072
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166052.3 |
NP_725387.3 |
1 - 2 |
267 |
62 |
View Full Product Details