Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scb
Entrez Gene ID
36692
Gene Name
scab
Gene Aliases
CG8095, CT21280, CT36929, Dm0620, Dmel\CG8095, Dmel_CG8095, E(Sev-CycE)E93, E(sev-cycE)[e93], ITA3, ItgaPS3, PS3, Vol, aPS3, alpha-PS3, alphaInt3, alphaPS3, alpha[[PS3]], beta[[PS3], l(2)01288, l(2)51Ea, vol
Chromosome Location
Chr.2R: 15248784 - 15258498 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0286785
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079026.3 NP_523750.2 7 - 8 3756 70
NM_166083.2 NP_725445.1 7 - 8 3417 70
GenBank mRNA BT015245.1 - 7 - 8 3739 70
BT021944.1 - 7 - 8 3389 70
U76605.1 - 7 - 8 3744 70

View Full Product Details