Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SK
Entrez Gene ID
31456
Gene Name
small conductance calcium-activated potassium channel
Gene Aliases
BcDNA:GM01744, CG10706, CG12733, CG4179, Dmel\CG10706, Dmel_CG10706, Sk, dSK
Chromosome Location
Chr.X: 5340236 - 5405138 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029761
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103415.2 NP_001096885.2 5 - 6 1222 94
NM_001272322.1 NP_001259251.1 8 - 9 2650 94
NM_001272323.2 NP_001259252.1 7 - 8 2548 94
NM_001272324.2 NP_001259253.1 7 - 8 2548 94
NM_001297957.1 NP_001284886.1 5 - 6 1467 94
NM_001297958.1 NP_001284887.1 7 - 8 2548 94
NM_001297959.1 NP_001284888.1 5 - 6 1461 94
NM_001297960.1 NP_001284889.1 7 - 8 2551 94
NM_080339.3 NP_525078.3 5 - 6 1461 94
NM_167029.3 NP_726986.3 7 - 8 2551 94
NM_167030.2 NP_726987.2 5 - 6 1461 94
NM_167031.3 NP_726988.3 7 - 8 2551 94
NM_167032.2 NP_726989.3 5 - 6 1467 94
NM_206631.4 NP_996354.2 7 - 8 2551 94

View Full Product Details