Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp28d2
Entrez Gene ID
33748
Gene Name
CG6081-PA
Gene Aliases
28d2, CG6081, Cyp28D2, Dmel\CG6081, Dmel_CG6081
Chromosome Location
Chr.2L: 5207267 - 5209345 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031688
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135067.3 NP_608911.1 3 - 4 486 66

View Full Product Details