Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cl
Entrez Gene ID
43938
Gene Name
clot
Gene Aliases
BcDNA:RE06767, CG11024, Dmel\CG11024, Dmel_CG11024
Chromosome Location
Chr.2L: 5520135 - 5521382 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000318
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273169.1 NP_001260098.1 3 - 4 436 84
NM_001273170.1 NP_001260099.1 3 - 4 421 84
NM_135086.4 NP_608930.1 2 - 3 362 84
GenBank mRNA AF503572.2 - 2 - 3 345 84
AY060849.1 - 2 - 3 350 84
AY113391.1 - 2 - 3 360 84
KX531732.1 - 2 - 3 221 84

View Full Product Details