The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001299053.1 | NP_001285982.1 | 2 - 3 | 806 | 125 |
NM_057908.4 | NP_477256.1 | 2 - 3 | 806 | 125 | |
NM_165157.2 | NP_723966.1 | 2 - 3 | 806 | 125 | |
NM_165158.1 | NP_723967.1 | 3 - 4 | 881 | 125 | |
GenBank mRNA | AY047561.1 | - | 2 - 3 | 787 | 125 |