Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dac
Entrez Gene ID
34982
Gene Name
dachshund
Gene Aliases
BG:DS02780.3, CG4952, DAC, Dac, Dac2-3, Dach, Dm-DAC, Dmel\CG4952, Dmel_CG4952, dach, l(2)36Ae, l(2)rK364, spn
Chromosome Location
Chr.2L: 16466512 - 16485998 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005677
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014486.3 NP_001014486.1 8 - 9 3254 63
NM_001014487.3 NP_001014487.1 9 - 10 3281 63
NM_165159.3 NP_723968.1 8 - 9 3260 63
NM_165160.3 NP_723969.1 8 - 9 3281 63
NM_165161.3 NP_723970.1 9 - 10 3287 63
NM_165162.3 NP_723971.1 9 - 10 3308 63

View Full Product Details