Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- eEF1delta
- Entrez Gene ID
-
34363
- Gene Name
- eukaryotic translation elongation factor 1 delta
- Gene Aliases
-
CG4912, Dmel\CG4912, Dmel_CG4912, eEF1d, eEf1delta
- Chromosome Location
-
Chr.2L: 10232507 - 10234450 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032198
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135517.4 |
NP_609361.1 |
1 - 2 |
153 |
82 |
View Full Product Details