Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vlc
Entrez Gene ID
46078
Gene Name
vulcan
Gene Aliases
CG8390, Dmel\CG8390, Dmel_CG8390, X, anon-41Fa, anon-WO0118547.98, l(2)07022
Chromosome Location
Chr.2R: 5702112 - 5705439 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259978
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169585.3 NP_001163056.1 4 - 5 1621 78
NM_001169586.1 NP_001163057.1 3 - 4 1536 78
NM_001259213.1 NP_001246142.1 4 - 5 1482 78
NM_001259214.2 NP_001246143.1 4 - 5 1364 78
NM_080129.5 NP_524868.1 4 - 5 1526 78
NM_165442.5 NP_724425.1 4 - 5 1449 78
NM_165443.4 NP_724426.3 4 - 5 1420 78
NM_165444.5 NP_724427.1 5 - 6 1383 78
GenBank mRNA AF275628.1 - 3 - 4 1355 78
AY061484.1 - 4 - 5 1505 78
BT100113.1 - 5 - 6 1383 78

View Full Product Details