Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aldh-III
Entrez Gene ID
45398
Gene Name
Aldehyde dehydrogenase type III
Gene Aliases
ALDH, ALDH-III, AldhIII, CG11140, CT41571, Dhap, Dmel\CG11140, Dmel_CG11140, FALDH, l(2)03610
Chromosome Location
Chr.2R: 7466879 - 7477162 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010548
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136441.4 NP_610285.5 3 - 4 556 67
NM_165528.3 NP_724562.2 3 - 4 1077 67
NM_165531.3 NP_724565.3 3 - 4 432 67
NM_165532.3 NP_724566.3 3 - 4 336 67
GenBank mRNA BT126071.1 - 3 - 4 583 67

View Full Product Details