Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nup44A
Entrez Gene ID
35762
Gene Name
Nucleoporin at 44A
Gene Aliases
CG8722, Dmel\CG8722, Dmel_CG8722, FBgn 33247, Nup44A/Seh1, SEH1, Seh1, nup44A, seh1
Chromosome Location
Chr.2R: 7988462 - 7991283 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033247
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136499.3 NP_610343.1 3 - 4 858 96
NM_165582.2 NP_724637.1 4 - 5 933 96
NM_165583.2 NP_724638.1 4 - 5 981 96
GenBank mRNA AY060480.1 - 3 - 4 2062 96

View Full Product Details